Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107186.1 | NP_001100656.1 | 12 - 13 | 2196 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Apoptosis Protein Kinase Detect Genomic DNA Rattus norvegicus Probe spans exons |