Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnni1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249865.2 | XP_006249927.1 | 1 - 2 | 106 | 84 | |
GenBank mRNA | FQ214889.1 | - | 1 - 2 | 84 | 84 | |
FQ215520.1 | - | 1 - 2 | 59 | 84 | ||
FQ215574.1 | - | 1 - 2 | 57 | 84 | ||
FQ215749.1 | - | 1 - 2 | 57 | 84 | ||
FQ216094.1 | - | 1 - 2 | 57 | 84 | ||
FQ216822.1 | - | 1 - 2 | 57 | 84 | ||
FQ217411.1 | - | 1 - 2 | 57 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |