Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcrla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100682.1 | NP_001094152.1 | 1 - 2 | 110 | 103 | |
GenBank mRNA | BC105902.1 | - | 1 - 2 | 81 | 103 | |
BC158820.1 | - | 1 - 2 | 110 | 7375319 | 103 | |
FQ225304.1 | - | 1 - 2 | 97 | 103 | ||
FQ227783.1 | - | 1 - 2 | 97 | 103 | ||
FQ232269.1 | - | 1 - 2 | 97 | 103 | ||
FQ233227.1 | - | 1 - 2 | 96 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |