Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199403.1 | NP_955435.1 | 13 - 14 | 1341 | 107 | |
XM_006250164.2 | XP_006250226.1 | 13 - 14 | 1796 | 107 | ||
XM_006250166.2 | XP_006250228.1 | 12 - 13 | 1775 | 107 | ||
XM_008769657.1 | XP_008767879.1 | 13 - 14 | 1808 | 107 | ||
XM_008769658.1 | XP_008767880.1 | 12 - 13 | 1787 | 107 | ||
XM_008769659.1 | XP_008767881.1 | 13 - 14 | 1808 | 107 | ||
GenBank mRNA | AJ421447.1 | - | 13 - 14 | 1341 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |