Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suco | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199403.1 | NP_955435.1 | 11 - 12 | 1264 | 71 | |
XM_006250164.2 | XP_006250226.1 | 11 - 12 | 1719 | 71 | ||
XM_008769657.1 | XP_008767879.1 | 11 - 12 | 1731 | 71 | ||
XM_008769659.1 | XP_008767881.1 | 11 - 12 | 1731 | 71 | ||
GenBank mRNA | AJ421447.1 | - | 11 - 12 | 1264 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |