Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107379.1 | NP_001100849.1 | 10 - 11 | 3107 | 99 | |
XM_006254719.2 | XP_006254781.1 | 10 - 11 | 3093 | 99 | ||
XM_006254720.2 | XP_006254782.1 | 10 - 11 | 2844 | 99 | ||
XM_006254721.2 | XP_006254783.1 | 10 - 11 | 2938 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |