Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004276.1 | NP_001004276.1 | 2 - 3 | 156 | 86 | |
XM_006254697.2 | XP_006254759.1 | 2 - 3 | 220 | 86 | ||
GenBank mRNA | BC079373.1 | - | 2 - 3 | 156 | 7110105 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |