Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013069.2 | NP_037201.1 | 5 - 6 | 579 | 106 | |
XM_006254761.2 | XP_006254823.1 | 5 - 6 | 637 | 106 | ||
GenBank mRNA | BC059152.1 | - | 5 - 6 | 560 | 6890709 | 106 |
FQ225098.1 | - | 5 - 6 | 554 | 106 | ||
FQ225666.1 | - | 3 - 4 | 198 | 106 | ||
FQ227357.1 | - | 5 - 6 | 556 | 106 | ||
FQ227455.1 | - | 5 - 6 | 559 | 106 | ||
FQ228116.1 | - | 5 - 6 | 562 | 106 | ||
FQ231756.1 | - | 5 - 6 | 106 | |||
FQ232412.1 | - | 5 - 6 | 604 | 106 | ||
FQ232423.1 | - | 5 - 6 | 593 | 106 | ||
FQ232867.1 | - | 5 - 6 | 563 | 106 | ||
FQ232996.1 | - | 5 - 6 | 556 | 106 | ||
FQ233568.1 | - | 5 - 6 | 562 | 106 | ||
FQ233706.1 | - | 5 - 6 | 562 | 106 | ||
FQ234457.1 | - | 5 - 6 | 565 | 106 | ||
X13044.1 | - | 5 - 6 | 535 | 106 | ||
X14254.1 | - | 5 - 6 | 543 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |