Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077667.1 | NP_001071135.1 | 4 - 5 | 451 | 89 | |
XM_008772113.1 | XP_008770335.1 | 5 - 6 | 745 | 89 | ||
XM_008772114.1 | XP_008770336.1 | 4 - 5 | 901 | 89 | ||
XM_008772115.1 | XP_008770337.1 | 4 - 5 | 524 | 89 | ||
XM_008772116.1 | XP_008770338.1 | 4 - 5 | 530 | 89 | ||
GenBank mRNA | BC127517.1 | - | 4 - 5 | 451 | 7104038 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |