Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191836.1 | NP_001178765.1 | 10 - 11 | 1312 | 75 | |
GenBank mRNA | S80376.1 | - | 10 - 11 | 1714 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |