Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106933.1 | NP_001100403.1 | 3 - 4 | 356 | 152 | |
XM_006254693.2 | XP_006254755.1 | 3 - 4 | 391 | 152 | ||
XM_006254694.2 | XP_006254756.1 | 3 - 4 | 389 | 152 | ||
XM_006254695.2 | XP_006254757.1 | 3 - 4 | 390 | 152 | ||
XM_006254696.2 | XP_006254758.1 | 3 - 4 | 390 | 152 | ||
XM_008772131.1 | XP_008770353.1 | 3 - 4 | 390 | 152 | ||
GenBank mRNA | BC167039.1 | - | 3 - 4 | 338 | 8368273 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |