Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iffo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108647.1 | NP_001102117.1 | 8 - 9 | 1601 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Rattus norvegicus Probe spans exons |