Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grcc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198725.1 | NP_001185654.1 | 1 - 2 | 350 | 100 | |
XM_006237325.2 | XP_006237387.1 | 1 - 2 | 371 | 100 | ||
GenBank mRNA | FM049692.1 | - | 1 - 2 | 350 | 100 | |
FM062242.1 | - | 1 - 2 | 129 | 100 |
Gene Symbol: LOC100911713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750659.3 | XP_003750707.1 | 1 - 2 | 372 | 100 | |
XM_006244638.2 | XP_006244700.1 | 1 - 2 | 377 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus Probe spans exons |