Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133512.2 | NP_598196.1 | 3 - 4 | 244 | 115 | |
XM_006237073.2 | XP_006237135.1 | 3 - 4 | 1426 | 115 | ||
XM_006237074.2 | XP_006237136.1 | 3 - 4 | 325 | 115 | ||
XM_006237075.2 | XP_006237137.1 | 3 - 4 | 394 | 115 | ||
XM_006237076.2 | XP_006237138.1 | 3 - 4 | 232 | 115 | ||
XM_006237077.2 | XP_006237139.1 | 3 - 4 | 282 | 115 | ||
GenBank mRNA | AF009511.1 | - | 2 - 3 | 198 | 115 | |
FQ241294.1 | - | 3 - 4 | 244 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |