Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763441.1 | XP_008761663.1 | 3 - 4 | 956 | 96 | |
XM_008763442.1 | XP_008761664.1 | 5 - 6 | 944 | 96 | ||
XM_008763443.1 | XP_008761665.1 | 6 - 7 | 1699 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Protein Kinase Rattus norvegicus Probe spans exons |