Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763441.1 | XP_008761663.1 | 5 - 6 | 1260 | 68 | |
XM_008763442.1 | XP_008761664.1 | 7 - 8 | 1248 | 68 | ||
XM_008763443.1 | XP_008761665.1 | 8 - 9 | 2003 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protein Kinase Rattus norvegicus Probe spans exons |