Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134555.1 | NP_001128027.1 | 7 - 8 | 1116 | 87 | |
GenBank mRNA | BC168673.1 | - | 7 - 8 | 1116 | 7937381 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |