Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017253.2 | NP_058949.1 | 2 - 3 | 247 | 128 | |
XM_008763370.1 | XP_008761592.1 | 2 - 3 | 260 | 128 | ||
XM_008763371.1 | XP_008761593.1 | 2 - 3 | 510 | 128 | ||
XM_008763372.1 | XP_008761594.1 | 3 - 4 | 336 | 128 | ||
XM_008763373.1 | XP_008761595.1 | 2 - 3 | 439 | 128 | ||
GenBank mRNA | AJ278701.1 | - | 2 - 3 | 241 | 128 | |
BC087710.1 | - | 2 - 3 | 247 | 7315507 | 128 | |
U35774.1 | - | 2 - 3 | 213 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |