Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024262.1 | NP_001019433.1 | 3 - 4 | 408 | 108 | |
XM_008763291.1 | XP_008761513.1 | 3 - 4 | 546 | 108 | ||
GenBank mRNA | BC092652.1 | - | 3 - 4 | 408 | 7320221 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |