Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itfg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108648.1 | NP_001102118.1 | 2 - 3 | 305 | 77 | |
XM_006237409.2 | XP_006237471.1 | 2 - 3 | 694 | 77 | ||
XM_006237410.2 | XP_006237472.1 | 2 - 3 | 695 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons |