Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191753.1 | NP_001178682.1 | 3 - 4 | 1149 | 104 | |
XM_006237659.2 | XP_006237721.1 | 2 - 3 | 1068 | 104 | ||
XM_006237660.2 | XP_006237722.1 | 3 - 4 | 1640 | 104 | ||
XM_008763381.1 | XP_008761603.1 | 3 - 4 | 3230 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |