Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053970.2 | NP_446422.2 | 9 - 10 | 1559 | 61 | |
XM_006231880.2 | XP_006231942.1 | 9 - 10 | 1713 | 61 | ||
XM_006231881.2 | XP_006231943.1 | 10 - 11 | 1791 | 61 | ||
GenBank mRNA | BC072687.1 | - | 9 - 10 | 1559 | 7099208 | 61 |
FQ228111.1 | - | 2 - 3 | 266 | 61 | ||
FQ230641.1 | - | 2 - 3 | 266 | 61 | ||
X87157.1 | - | 9 - 10 | 1667 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |