Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankdd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760686.1 | XP_008758908.1 | 10 - 11 | 1151 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus Probe spans exons |