Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207595.1 | NP_997478.1 | 4 - 5 | 722 | 145 | |
XM_006257186.2 | XP_006257248.1 | 3 - 4 | 528 | 145 | ||
GenBank mRNA | AY566568.1 | - | 4 - 5 | 722 | 145 | |
BC081975.1 | - | 4 - 5 | 911 | 7126874 | 145 |
Gene Symbol: LOC100910438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760690.1 | XP_008758912.1 | 4 - 5 | 974 | 145 | |
XM_008760691.1 | XP_008758913.1 | 3 - 4 | 528 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |