Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002728994.4 | XP_002729040.2 | 3 - 4 | 765 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |