Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011946.1 | NP_001011946.1 | 681 | 76 | ||
GenBank mRNA | BC079376.1 | - | 681 | 7110247 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |