Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008760700.1 | XP_008758922.1 | 8 - 9 | 762 | 129 | |
GenBank mRNA | X83579.1 | - | 8 - 9 | 608 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Rattus norvegicus Probe spans exons |