Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031821.1 | NP_114009.1 | 11 - 12 | 1743 | 69 | |
GenBank mRNA | AF136583.1 | - | 11 - 12 | 1743 | 69 | |
BC070878.1 | - | 11 - 12 | 1727 | 7096750 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |