Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231951.2 | XP_006232013.1 | 4 - 5 | 2832 | 66 | |
XM_008760741.1 | XP_008758963.1 | 3 - 4 | 369 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons |