Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108710.1 | NP_001102180.1 | 3 - 4 | 154 | 110 | |
GenBank mRNA | BC160874.1 | - | 125 | 7367398 | 110 | |
FQ221314.1 | - | 211 | 183 |
Set Membership: |
Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Rattus norvegicus Probe spans exons |