Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tiparp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003749294.3 | XP_003749342.1 | 2 - 3 | 1218 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons |