Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100647.1 | NP_001094117.1 | 7 - 8 | 679 | 80 | |
GenBank mRNA | BC160929.1 | - | 7 - 8 | 679 | 8364923 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Rattus norvegicus Probe spans exons |