Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033681.1 | NP_001028853.1 | 2 - 3 | 274 | 137 | |
XM_006246635.2 | XP_006246697.1 | 3 - 4 | 504 | 137 | ||
XM_006246637.2 | XP_006246699.1 | 3 - 4 | 472 | 137 | ||
XM_006246638.1 | XP_006246700.1 | 2 - 3 | 493 | 137 | ||
GenBank mRNA | BC101891.1 | - | 2 - 3 | 274 | 7930704 | 137 |
FQ212627.1 | - | 1 - 2 | 89 | 137 | ||
FQ213502.1 | - | 2 - 3 | 311 | 137 | ||
FQ219881.1 | - | 2 - 3 | 137 | |||
FQ220539.1 | - | 2 - 3 | 288 | 137 | ||
FQ226884.1 | - | 2 - 3 | 286 | 137 | ||
FQ229255.1 | - | 2 - 3 | 297 | 137 | ||
FQ231501.1 | - | 2 - 3 | 256 | 137 | ||
FQ232467.1 | - | 2 - 3 | 256 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |