Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105775.2 | NP_001099245.2 | 421 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |