Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034152.1 | NP_001029324.1 | 3 - 4 | 352 | 55 | |
GenBank mRNA | BC105916.1 | - | 3 - 4 | 352 | 7384671 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |