Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133580.1 | NP_598264.1 | 3 - 4 | 242 | 78 | |
GenBank mRNA | BC061984.1 | - | 4 - 5 | 550 | 5599180 | 78 |
U18771.1 | - | 3 - 4 | 242 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |