Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109443.1 | NP_001102913.1 | 2 - 3 | 382 | 96 | |
GenBank mRNA | FQ218220.1 | - | 2 - 3 | 327 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |