Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wsb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042561.1 | NP_001036026.1 | 1 - 2 | 134 | 114 | |
GenBank mRNA | BC167746.1 | - | 1 - 2 | 137 | 8364583 | 114 |
DY312839.1 | - | 1 - 2 | 136 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |