Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013116.2 | NP_001013134.2 | 8 - 9 | 1634 | 68 | |
XM_006246703.2 | XP_006246765.1 | 8 - 9 | 1683 | 68 | ||
XM_008767826.1 | XP_008766048.1 | 8 - 9 | 1772 | 68 | ||
GenBank mRNA | BC101901.1 | - | 7 - 8 | 1459 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |