Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sqstm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175843.4 | NP_787037.2 | 2 - 3 | 330 | 154 | |
NM_181550.1 | NP_853528.1 | 2 - 3 | 296 | 154 | ||
XM_006246213.2 | XP_006246275.1 | 2 - 3 | 352 | 154 | ||
XM_006246214.2 | XP_006246276.1 | 2 - 3 | 352 | 154 | ||
XM_006246215.2 | XP_006246277.1 | 2 - 3 | 352 | 154 | ||
GenBank mRNA | AF439403.1 | - | 2 - 3 | 296 | 154 | |
BC061575.1 | - | 2 - 3 | 334 | 6887571 | 154 | |
Y08355.1 | - | 2 - 3 | 359 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |