Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013972.2 | NP_001013994.1 | 5 - 6 | 678 | 122 | |
GenBank mRNA | BC081930.1 | - | 5 - 6 | 567 | 122 | |
BC127522.1 | - | 5 - 6 | 678 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |