Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005873.1 | NP_001005873.1 | 15 - 16 | 1997 | 116 | |
GenBank mRNA | BC083821.1 | - | 15 - 16 | 1997 | 7111216 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |