Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006602.1 | NP_001006602.1 | 7 - 8 | 821 | 59 | |
GenBank mRNA | BC083862.1 | - | 7 - 8 | 821 | 7126912 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons |