Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031702.1 | NP_113890.1 | 4 - 5 | 775 | 97 | |
XM_006246805.2 | XP_006246867.1 | 3 - 4 | 975 | 97 | ||
GenBank mRNA | AJ011811.1 | - | 3 - 4 | 411 | 97 | |
BC101892.1 | - | 4 - 5 | 775 | 7930797 | 97 | |
BC159404.1 | - | 3 - 4 | 869 | 7458165 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |