Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013118.1 | NP_001013136.1 | 5 - 6 | 941 | 59 | |
XM_008768045.1 | XP_008766267.1 | 5 - 6 | 683 | 59 | ||
GenBank mRNA | BC079102.1 | - | 5 - 6 | 941 | 7124908 | 59 |
FQ225352.1 | - | 5 - 6 | 671 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons |