Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aipl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021590.1 | NP_067601.1 | 5 - 6 | 847 | 67 | |
XM_006246798.2 | XP_006246860.1 | 5 - 6 | 879 | 67 | ||
GenBank mRNA | AF180340.1 | - | 5 - 6 | 847 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Rattus norvegicus 1700 Probe spans exons |