Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022920.1 | NP_075209.1 | 7 - 8 | 1646 | 98 | |
XM_006246225.2 | XP_006246287.1 | 7 - 8 | 1785 | 98 | ||
GenBank mRNA | D13963.1 | - | 7 - 8 | 1646 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Rattus norvegicus Probe spans exons |