Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc47a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014118.2 | NP_001014140.1 | 5 - 6 | 496 | 91 | |
GenBank mRNA | AB248823.1 | - | 5 - 6 | 496 | 91 | |
AB248824.1 | - | 5 - 6 | 496 | 91 | ||
BC088413.1 | - | 516 | 7109897 | 91 | ||
FQ217802.1 | - | 510 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons |