Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006247233.2 | XP_006247295.1 | 4 - 5 | 698 | 61 | |
XM_006247234.2 | XP_006247296.1 | 4 - 5 | 702 | 61 | ||
XM_006247235.2 | XP_006247297.1 | 4 - 5 | 673 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |