Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017067.1 | NP_058763.1 | 1 - 2 | 50 | 65 | |
GenBank mRNA | L13722.1 | - | 1 - 2 | 50 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |